Skip to Content
Merck
Bupropion Hydrobromide pharmaceutical secondary standard, certified reference material

Bupropion hydrobromide

Bupropion Hydrobromide pharmaceutical secondary standard, certified reference material
Synonym(s):
(2RS)-1-(3-Chlorophenyl)-2-[(1,1-dimethylethyl)amino]propan-1-one hydrobromide, Bupropion hydrobromide
Empirical Formula (Hill Notation):
C13H18ClNO · HBr
CAS Number:
Molecular Weight:
320.65

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade