Skip to Content
Merck
Boc-Inp-OH ≥99.0% (HPLC)

Boc-Inp-OH

Boc-Inp-OH ≥99.0% (HPLC)
Synonym(s):
1-Boc-piperidine-4-carboxylic acid, Boc-isonipecotic acid
Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing