Benzyl chloride
Synonym(s):
α-Chlorotoluene, Benzyl chloride
Linear Formula:
C6H5CH2Cl
CAS Number:
Molecular Weight:
126.58
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing