
Atranorin

Synonym(s):
3-Formyl-2,4-dihydroxy-6-methylbenzoic acid 3-hydroxy-4-(methoxycarbonyl)-2,5-dimethylphenyl ester, Atranoric acid, Parmelin, Parmelin acid, Usnarin, Usnarin acid
Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing