α-Santonin
Synonym(s):
(−)-α-Santonin, (3S,5aS,9bS)-3a,5,5a,9b-Tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)dione, Semenen
Empirical Formula (Hill Notation):
C15H18O3
CAS Number:
Molecular Weight:
246.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1