
Anti-EP300 antibody produced in rabbit

Synonym(s):
Anti-E1A-associated protein p300, Anti-HAT p300, Anti-Histone acetyltransferase p300
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing