
7-Ethoxy-4-(trifluoromethyl)coumarin

Synonym(s):
Ethyl 4-(trifluoromethyl)umbelliferyl ether
Empirical Formula (Hill Notation):
C12H9F3O3
CAS Number:
Molecular Weight:
258.19
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing