
4,5-Dimethoxy-2-nitrobenzyl chloroformate

Synonym(s):
6-Nitroveratryl chloroformate, 6-Nitroveratryloxycarbonyl chloride, NVOC chloride, NVOC-Cl
Linear Formula:
ClCO2CH2C6H2(OCH3)2NO2
CAS Number:
Molecular Weight:
275.64
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing