
3-(Trimethylsilyl)propynoic acid

Synonym(s):
3-(Trimethylsilyl)propiolic acid
Linear Formula:
(CH3)3SiC≡CCO2H
CAS Number:
Molecular Weight:
142.23
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing