Skip to Content
BI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Phenyl-1-propanol
Synonym(s):
3-Phenylpropyl alcohol, Hydrocinnamyl alcohol
Linear Formula:
C
6
H
5
(CH
2
)
3
OH
CAS Number:
122-97-4
Molecular Weight:
136.19
EC Number:
204-587-6
Products
Product Category
aroma chemicals (1)
building blocks (1)
Brand
Sigma-Aldrich (1)
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
140856
3-Phenyl-1-propanol,
98%
Expand
View Pricing
W288500
3-Phenyl-1-propanol,
≥98%, FCC
Expand
View Pricing