Skip to Content
Merck
(3-Mercaptopropyl)methyldimethoxysilane ≥95.0%

(3-Mercaptopropyl)methyldimethoxysilane

(3-Mercaptopropyl)methyldimethoxysilane ≥95.0%
Synonym(s):
Dimethoxy-(3-mercaptopropyl)methylsilane
Linear Formula:
CH3Si(OCH3)2CH2CH2CH2SH
CAS Number:
Molecular Weight:
180.34
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing