
3-Hydroxy-DL-kynurenine

Synonym(s):
α,2-Diamino-3-hydroxy-γ-oxobenzenebutanoic acid
Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing