
3-Dehydroquinic acid potassium salt

Synonym(s):
(1R,3R,4S)-1,3,4-Trihydroxy-5-oxocyclohexanecarboxylic acid potassium salt, 5-Dehydroquinic acid potassium salt
Empirical Formula (Hill Notation):
C7H9KO6
CAS Number:
Molecular Weight:
228.24
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing