
2-Bromothioanisole

Synonym(s):
2-Bromophenyl methyl sulfide
Linear Formula:
BrC6H4SCH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing