Skip to Content
Merck
All Photos(1)

Key Documents

PHL89537

Procyanidin C1

phyproof® Reference Substance

Synonym(s):

Cinnamtannin A1, Proanthocyanidin C1, Procyanidol C1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H38O18
CAS Number:
Molecular Weight:
866.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4c([C@@H]5[C@@H](O)[C@H](Oc6cc(O)cc(O)c56)c7ccc(O)c(O)c7)c(O)cc(O)c34)c8ccc(O)c(O)c8)c2O[C@@H]1c9ccc(O)c(O)c9

InChI

1S/C45H38O18/c46-18-10-27(54)33-32(11-18)61-42(16-2-5-21(48)25(52)8-16)39(59)37(33)35-29(56)14-30(57)36-38(40(60)43(63-45(35)36)17-3-6-22(49)26(53)9-17)34-28(55)13-23(50)19-12-31(58)41(62-44(19)34)15-1-4-20(47)24(51)7-15/h1-11,13-14,31,37-43,46-60H,12H2/t31-,37-,38+,39-,40-,41-,42-,43-/m1/s1

InChI key

MOJZMWJRUKIQGL-XILRTYJMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Nanofunctionalization of Additively Manufactured Titanium Substrates for Surface-Enhanced Raman Spectroscopy Measurements.: This article explores the functionalization of titanium substrates with nanomaterials for use in surface-enhanced Raman spectroscopy (SERS). Pyridine is utilized as a probe molecule to enhance the detection sensitivity of the SERS technique (Pisarek M et al., 2022).

  • UPLC-MS/MS determination of steroid hormones via a novel reaction based on derivatisation by a cyclic-organophosphate.: This research introduces a new derivatization method involving pyridine for the ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) determination of steroid hormones, significantly enhancing analytical performance (Szterk A et al., 2019).



Other Notes

This compound is commonly found in plants of the genus: cinnamomum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service