Skip to Content
Merck
All Photos(1)

Documents

442599

Supelco

γ-Chlordane

PESTANAL®, analytical standard

Synonym(s):

trans-Chlordane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Cl8
CAS Number:
Molecular Weight:
409.78
EC Number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)[C@@]2(Cl)[C@](C[C@@H](Cl)[C@@H]3Cl)([H])[C@]3([H])[C@]1(Cl)C2(Cl)Cl

InChI

1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2-,3+,4-,5-,8-,9+/m0/s1

InChI key

BIWJNBZANLAXMG-MPPHZFHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Chlordane belongs to the class of organochlorine pesticides (OCPs) typically utilized for agriculture applications.

Application

γ-Chlordane may be used as an analytical reference standard for the determination of the analyte in meat samples, marine shellfishes, vegetation samples and water samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a novel method for the analysis of chlorinated pesticides in soils using microwave-assisted extraction-headspace solid phase microextraction and gas chromatography-tandem mass spectrometry.
Herbert p, et al.
Analytical and Bioanalytical Chemistry, 384(3), 810-816 (2006)
Multiresidue analysis of organochlorine and pyrethroid pesticides in ground beef meat by gas chromatography-mass spectrometry.
Stefanelli P, et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(4), 350-356 (2009)
Multiresidual analysis of organochlorine pesticides, polychlorinated biphenyls and polycyclic aromatic hydrocarbons in marine shellfishes by gas chromatography-ion trap mass spectrometry.
Jie F and Kai-Xiong W
Chinese Journal of Analytical Chemistry, 35(11), 1607-1613 (2007)
Microwave-assisted extraction versus Soxhlet extraction in the analysis of 21 organochlorine pesticides in plants.
Barriada-Pereira M, et al.
Journal of Chromatography A, 1008(1), 115-122 (2003)
D de Almeida Azevedo et al.
Journal of chromatography. A, 879(1), 13-26 (2000-06-28)
Gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS) were optimized and applied for the trace-level determination of 42 priority pesticides and 33 priority organic pollutants from European Union Directive EC 76/464. First, off-line solid-phase extraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service