Skip to Content
Merck
All Photos(1)

Key Documents

Z614610

Vivaspin 20 centrifugal concentrator

MWCO 10 kDa

Synonym(s):

Vivaspin 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

material

polycarbonate housing
polyethersulfone membrane

description

PES membrane

sterility

non-sterile

feature

holdup volume 20 μL

packaging

package of 48 ea

manufacturer/tradename

Sartorius VS2002

parameter

5 bar max. pressure
8000 × g max. centrifugal force

dead stop volume

50 μL

membrane area

6 cm2

tube size

~50 mL

working volume

20 mL

pore size

10 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

The Vivaspin 20 mL centrifugal concentrator has been developed to offer increased volume flexibility and performance. Vivaspin 20 can be used in either swing bucket of fixed angle rotors (carriers accepting 15 mL and 50 mL bottles). Vivaspin features twin vertical membranes for unparalleled filtration speed and 100X plus concentrations.

Remaining volume is easy to read off the printed scales on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recover of the final concentrate.

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin

Note: One step dealing of concentrated sample using diafiltration cup. Gas pressure filtration; for faster processing gas pressure can be combined with centrifugal force.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service