Skip to Content
Merck
All Photos(1)

Key Documents

SML2899

Sigma-Aldrich

Humanin Trifluoroacetate

≥95% (HPLC)

Synonym(s):

L-Methionyl-L-alanyl-L-prolyl-L-arginylglycyl-L-phenylalanyl-L-seryl-L-cysteinyl-L-leucyl-L-leucyl-L-leucyl-L-leucyl-L-threonyl-L-seryl-L-?-glutamyl-L-isoleucyl-L-?-aspartyl-L-leucyl-L-prolyl-L-valyl-L-lysyl-L-arginyl-L-arginyl-L-alanine trifluoroacetate, MAPRGFSCLLLLTSEIDLPVKRRA trifluoroacetate, Met-Ala-Pro-Arg-Gly-Phe-Ser-Cys-Leu-Leu-Leu-Leu-Thr-Ser-Glu-Ile-Asp-Leu-Pro-Val-Lys-Arg-Arg-Ala

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C119H204N34O32S2 · xC2HF3O2
Molecular Weight:
2687.23 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to beige

storage temp.

−20°C

Biochem/physiol Actions

Humanin is 24-amino acid mitochondria-associated peptide that is involved in numerous biological processes including apoptosis, cell survival, substrate metabolism, inflammatory response, and response to stressors such as oxidative stress, ischemia, and starvation. Humanin exhibits protective effects against cellular apoptosis. Apparently it inhibits BAX translocation to the mitochondrial outer membrane.
mitochondria-associated peptide; exhibits protective effects against cellular apoptosis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Cristina Zárate et al.
Frontiers in aging neuroscience, 11, 123-123 (2019-06-20)
Astroglial cells are crucial for central nervous system (CNS) homeostasis. They undergo complex morpho-functional changes during aging and in response to hormonal milieu. Ovarian hormones positively affect different astroglia parameters, including regulation of cell morphology and release of neurotrophic and
Daniel L Morris et al.
The Journal of biological chemistry, 294(50), 19055-19065 (2019-11-07)
The mitochondrial, or intrinsic, apoptosis pathway is regulated mainly by members of the B-cell lymphoma 2 (BCL-2) protein family. BCL-2-associated X apoptosis regulator (BAX) plays a pivotal role in the initiation of mitochondria-mediated apoptosis as one of the factors causing
Y Hashimoto et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(11), 6336-6341 (2001-05-24)
Through functional expression screening, we identified a gene, designated Humanin (HN) cDNA, which encodes a short polypeptide and abolishes death of neuronal cells caused by multiple different types of familial Alzheimer's disease genes and by Abeta amyloid, without effect on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service