Skip to Content
Merck
All Photos(2)

Key Documents

SMB00424

Sigma-Aldrich

Platycodin D

≥98% (HPLC)

Synonym(s):

3β-(β-D-glucopyranosyloxy)-2β,16α,23,24-tetrahydroxy-O-D-apio-β-D-furanosyl-(1→3)-O-β-D-xylopyranosyl-(1→4)-O-6-deoxy-olean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H92O28
CAS Number:
Molecular Weight:
1225.32
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)C(CO)(CO)[C@]3([H])CC[C@@]4(C)[C@]5(C)C[C@@H](O)[C@@]6(C(O[C@H]7[C@H](O[C@@H]8O[C@@H](C)[C@H](O[C@H]9[C@H](O)[C@@H](O[C@@H]%10OC[C@@](CO)(O)[C@H]%10O)[C@H](O)CO9)[C@@H](O)[C@H]8O)[C@@H](O)[C@@H](O

InChI

1S/C57H92O28/c1-23-40(81-45-39(72)41(30(64)18-76-45)82-49-43(73)56(75,21-61)22-78-49)36(69)38(71)46(79-23)83-42-33(66)29(63)17-77-48(42)85-50(74)57-12-11-51(2,3)14-27(57)24-7-9-52(4)26-13-28(62)44(84-47-37(70)35(68)34(67)31(16-58)80-47)55(19-59,20-60)25(26)8-10-54(52,6)53(24,5)15-32(57)65/h7,23,25-49,58-73,75H,8-22H2,1-6H3/t23-,25+,26-,27-,28-,29-,30+,31+,32+,33-,34+,35-,36-,37+,38+,39+,40-,41-,42+,43-,44-,45-,46-,47-,48?,49-,52+,53+,54+,56+,57+/m0/s1

InChI key

RJDYKSRQTBCKLC-XTIQGCOGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Platycodin D (PD) is a triterpenoid saponin found in the root of Platycodon grandifloras called Platycodonis Radix.

Application

Platycodin D has been used:
  • to test its anti-inflammatory and inhibitory effect on eosinophilic airway inflammation and T-helper type 2 (Th2) signaling pathways using ovalbumin (OVA)-induced allergic asthma mouse model
  • to test its anti-obese effects on the activation of brown adipose tissue featuring in vivo and in vitro studies
  • to test its efficacy as an adjuvant with infectious bronchitis (IB) vaccine in chicken

Biochem/physiol Actions

Platycodin D elicits strong anti-cancer properties and exhibits cytotoxicity against several cancer cell lines of both in vitro and in vivo origin. It exerts anti-inflammatory and protective effects against lipopolysaccharide (LPS)-induced acute lung injury (ALI) by upregulating liver-X receptors α (LXRα) and ATP-binding cassette transporter A1 (ABCA1) signaling pathway. It also possesses protective effects against Cisplatin (CDDP)-induced nephrotoxicity in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae-Won Kim et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(12), 4254-4259 (2012-05-24)
Platycodin D (PD) is well known as a potent triterpenoid saponin having various pharmacological activities isolated from the root of Platycodon grandiflorum (Jacq.) A. DC. (Campanulaceae). We aimed to evaluate protective effect of PD on cisplatin (CDDP)-induced nephrotoxicity. Male ICR
Yefei Zhou et al.
The journal of poultry science, 57(2), 160-167 (2020-05-29)
Adjuvants are common vaccine components. Novel adjuvants may improve the protective immunity conferred by vaccines against poultry diseases. Here, a less-hemolytic saponin, platycodin D (PD), isolated from the root of Platycodon grandiflorum was investigated as a potential alternative adjuvant. PD
Eung Gu Lee et al.
The Journal of asthma : official journal of the Association for the Care of Asthma, 1-11 (2021-06-16)
Introduction: Bronchial asthma is a common chronic inflammatory condition of the airway tissue. Platycodin D (PLD) has antiinflammatory effects in a mouse model of allergic asthma. In this work, the anti-asthma potential of PLD was studied by investigation of its
Yuan-Chuan Lin et al.
The American journal of Chinese medicine, 46(3), 537-549 (2018-03-30)
Platycodin D (PD) is the main active saponin isolated from Platycodon grandiflorum (PG) and is reported to exhibit anticancer, anti-angiogenic, anti-inflammation and anti-obesity biological effects. The current study aims to evaluate the therapeutic efficacy of PD in cardiac fibrosis and
Muhammad Khan et al.
Journal of cellular and molecular medicine, 20(3), 389-402 (2015-12-10)
Cancer is a multi-faceted disease comprised of a combination of genetic, epigenetic, metabolic and signalling aberrations which severely disrupt the normal homoeostasis of cell growth and death. Rational developments of highly selective drugs which specifically block only one of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service