Skip to Content
Merck
All Photos(4)

Key Documents

SAB4504104

Sigma-Aldrich

Anti-phospho-PKD2 (pSer876) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HSPC187

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 96 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:60000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer876)

Gene Information

human ... PRKD2(25865)

General description

The protein kinase D2 (PRKD2) gene, mapped to human chromosome 4q21-23, codes for integral membrane protein polycystin-2. PRKD2 is a member of the serine/threonine protein kinase D (PRKD) family. It is characterized with two N-terminal cysteine-rich zinc finger domains involved in binding diacylglycerol (DAG) and phorbol esters, a central lipid/protein binding pleckstrin homology (PH) domain, and a C-terminal kinase domain. PRKD2 is mainly expressed in the endoplasmic reticulum (ER).

Immunogen

The antiserum was produced against synthesized peptide derived from human PKD2 around the phosphorylation site of Ser876.

Immunogen Range: 829-878

Biochem/physiol Actions

Protein kinase D2 (PRKD2) is a novel, essential mediator of tumor cell-endothelial cell communication and a promising therapeutic target to inhibit angiogenesis in gastrointestinal cancers. Mutation in the gene leads to autosomal dominant polycystic kidney disease (ADPKD). Polycystin-2 might function as a component of intracellular channel complexes. PRKD2 influences CD8+ T cell-dependent immune responses. PKD2 facilitates cell migration and it is associated with the progression and maintenance of various malignancies. Increased expression of PKD2 is observed in various types of cancers including colorectal cancer, glioblastoma multiforme (GBM). CRT0066101 acts as an inhibitor for all the three protein kinase D isoforms. Therefore, it can be used as a therapeutic drug for PRKD2 induced carcinogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nisha Durand et al.
Journal of clinical medicine, 5(2) (2016-02-06)
The Protein Kinase D (PKD) isoforms PKD1, PKD2, and PKD3 are effectors of the novel Protein Kinase Cs (nPKCs) and diacylglycerol (DAG). PKDs impact diverse biological processes like protein transport, cell migration, proliferation, epithelial to mesenchymal transition (EMT) and apoptosis.
Y Cai et al.
The Journal of biological chemistry, 274(40), 28557-28565 (1999-09-25)
PKD2, the second gene for the autosomal dominant polycystic kidney disease (ADPKD), encodes a protein, polycystin-2, with predicted structural similarity to cation channel subunits. However, the function of polycystin-2 remains unknown. We used polyclonal antisera specific for the intracellular NH(2)
T Mochizuki et al.
Science (New York, N.Y.), 272(5266), 1339-1342 (1996-05-31)
A second gene for autosomal dominant polycystic kidney disease was identified by positional cloning. Nonsense mutations in this gene (PKD2) segregated with the disease in three PKD2 families. The predicted 968-amino acid sequence of the PKD2 gene product has six
Ninel Azoitei et al.
Gut, 59(10), 1316-1330 (2010-08-25)
Tumour angiogenesis is crucially dependent on the communication between the tumour and the associated endothelium. Protein kinase D (PKD) isoenzymes mediate vascular endothelial growth factor-A (VEGF-A) induced endothelial cell proliferation and migration and are also highly expressed in various tumours.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service