BR782970
BRAND® Insert strips 9mm for 24 x 6 well BRANDplates®
smooth-walled, 4 Inserts/Strip, PET membrane, pore size 8 μm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
Recommended Products
material
PET membrane
description
cellGrade(TM) plus
sterility
sterile
packaging
pack of 1 ea (12 strips with 4 inserts per strip)
manufacturer/tradename
BRAND 782970
size
6 well
pore size
8 μm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service