Skip to Content
Merck
All Photos(1)

Key Documents

92396

Sigma-Aldrich

Fluorescein di-(β-D-glucopyranoside)

Synonym(s):

FDGlu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32O15
CAS Number:
Molecular Weight:
656.59
Beilstein:
7455945
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥85.0% (HPLC)

form

solid

fluorescence

λex 490 nm; λem 514 nm±10 nm in H2O

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3c(Oc4cc(O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)ccc4C36OC(=O)c7ccccc67)c2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C32H32O15/c33-11-21-23(35)25(37)27(39)30(45-21)42-13-5-7-17-19(9-13)44-20-10-14(43-31-28(40)26(38)24(36)22(12-34)46-31)6-8-18(20)32(17)16-4-2-1-3-15(16)29(41)47-32/h1-10,21-28,30-31,33-40H,11-12H2/t21-,22-,23-,24-,25+,26+,27-,28-,30-,31-/m1/s1

InChI key

ZTOBILYWTYHOJB-LWXMCBIJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service