Skip to Content
Merck
All Photos(1)

Key Documents

W338020

Sigma-Aldrich

Linoleic acid

≥95%, FG

Synonym(s):

cis-9,cis-12-Octadecadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
Beilstein:
1727101
EC Number:
Council of Europe no.:
694
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
faint; fatty
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 184.1065

Assay

≥95%

refractive index

n20/D 1.466 (lit.)

bp

229-230 °C/16 mmHg (lit.)

mp

−5 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

faint; fatty

storage temp.

2-8°C

SMILES string

OC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes [1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service