Skip to Content
Merck
All Photos(2)

Key Documents

NIST2891

Ethanol-water solution

NIST® SRM® 2891, nominal mass fraction 0.02%

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material is a solution of ethanol in water with a nominal concentration of 0.02 % by mass. A single unit of SRM 2891 contains five 2-milliliter ampoules, each with an approximate 1.2 mL of the solution. For more information, please refer to the SDS and COA.
SRM 2891_cert
SRM 2891_SDS

Application

Ethanol-water solution is intended for use with calibrating instruments and techniques used for the determination of ethanol in blood.

Features and Benefits

  • The certified values for ethanol are provided, that are metrological traceability to the International System of Units (SI) derived unit for mass fraction, expressed as a percentage and determined by gas chromatography with flame ionization detection (GC-FID) and Karl Fischer analysis.
  • Expiration details and instructions for use, storage and handling are provided on the NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Ethanol (C2H6O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service