Skip to Content
Merck
All Photos(1)

Key Documents

NIST1763B

Low Alloy Steel

disk form, NIST® SRM® 1763b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

disc

packaging

pkg of 1 discs

manufacturer/tradename

NIST®

General description

Each unit of SRM 1763b contains a disk with a diameter of about 34 mm and a thickness of 19 mm.



SRM 1763B_cert

SRM 1763B_SDS

Application

Low alloy steel standard reference material (SRM) is intended for assessing methods employed inanalyzing elements within low alloy steel and materials with a similar matrix.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of elements in a steel matrix.
  • Information value for oxygen is reported as mass fractions.
  • The certified values are metrologically traceable to the SI derived unit of mass fraction, expressed as a percent.
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Antimony (Sb), Arsenic (As), Boron (B), Carbon (C), Cobalt (Co), Chromium (Cr), Copper (Cu), Manganese (Mn), Molybdenum (Mo), Niobium (Nb), Nickel (Ni), Phosphorus (P), Silicon (Si), Sulfur (S), Tantalum (Ta), Tin (Sn), Titanium (Ti), Tungsten (W), Vanadium (V), Zirconium (Zr)

Reference/Informational value is provided for the following analytes.Iron (Fe)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service