Skip to Content
Merck
All Photos(1)

Documents

N2260

Supelco

D(−)-Norgestrel

analytical standard

Synonym(s):

13β-Ethyl-17α-ethynyl-17β-hydroxygon-4-en-3-one, 17α-Ethynyl-18-homo-19-nor-testosterone, 18,19-Dinor-13β-ethyl-17β-hydroxy-4-pregnen-20-yn-3-one, 18,19-Dinor-4-pregnen-20-yn-3-one, Levonorgestrel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

InChI

1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

Gene Information

human ... PGR(5241)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

D(−)-Norgestrel is a hormonal medication and a contraceptive used by women.

Application

D(−)-Norgestrel may be used as a reference standard for the analysis of the analyte in human plasma using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Methods for Therapeutic Drug Monitoring and Toxicology (2011)
Anita Nelson et al.
Obstetrics and gynecology, 122(6), 1205-1213 (2013-11-19)
To evaluate the efficacy and safety of two low-dose levonorgestrel intrauterine contraceptive systems. Nulliparous and parous women aged 18-35 years with regular menstrual cycles (21-35 days) requesting contraception were randomized to 3 years of treatment with one of two levonorgestrel
Alice W Y Wong et al.
Obstetrics and gynecology, 121(5), 943-950 (2013-05-03)
To estimate the rate of endometrial pathology with the prophylactic use of levonorgestrel-releasing intrauterine system in women with breast cancer treated with tamoxifen. This was a randomized contro-lled trial of 129 Chinese women who attended a university hospital in Hong
Satu Heliövaara-Peippo et al.
American journal of obstetrics and gynecology, 209(6), 535-535 (2013-09-04)
Menorrhagia is a common problem impairing the quality of life (QOL) of many women. Both levonorgestrel-releasing intrauterine system (LNG-IUS) and hysterectomy are effective treatment modalities but no long-term comparative studies of QOL and costs exist. The objective of this study
Elizabeth A McCarthy et al.
Contraception, 86(5), 587-590 (2012-04-14)
Correctly placed levonorgestrel-releasing intrauterine systems (LNG-IUSs) are rarely associated with intrauterine pregnancy when pregnancy occurs. LNG-IUS retrieval, termination of pregnancy and conservative management if retracted strings prevent ready removal are the usual clinical options given to women. The conservative course

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service