Skip to Content
Merck
All Photos(1)

Key Documents

52445

Supelco

Anthrone

analytical standard

Synonym(s):

9(10H)-Anthracenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
Beilstein:
1910173
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

154-157 °C (lit.)

suitability

conforms to structure for Infrared spectrum

format

neat

storage temp.

2-8°C

SMILES string

O=C1c2ccccc2Cc3ccccc13

InChI

1S/C14H10O/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-8H,9H2

InChI key

RJGDLRCDCYRQOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giorgia Vici et al.
Foods (Basel, Switzerland), 10(6) (2021-07-03)
Gluten-free diets are often characterized by an inadequate intake of nutrients and are generally monotonous for the limited number of products celiac patients can use. As rice is the most used cereal by celiac consumers, studying rice varieties nutritional characteristics
Yasuaki Arai et al.
Asia-Pacific journal of clinical oncology, 11(1), 41-48 (2015-01-30)
This prospective multicenter study aimed to evaluate the efficacy and adverse events of hepatic arterial infusion chemotherapy (HAIC) using percutaneous catheter placement techniques for liver metastases from colorectal cancer (CRC). We administered 5-fluorouracil at 1000 mg/m2 over 5 h via hepatic arterial
Yoichiro Kato et al.
AoB PLANTS, 6 (2014-10-02)
Floods are major constraints to crop production worldwide. In low-lying, flood-prone areas of the tropics, longer-term partial submergence (stagnant flooding [SF]) greatly reduces rice yield. This study assesses shoot growth and several physiological mechanisms associated with SF tolerance in rice.
Xia Chen et al.
Carbohydrate polymers, 112, 469-474 (2014-08-19)
A protein-containing polysaccharide, EPS2BW, was fractionated from the exopolysaccharide (EPS) produced by a medicinal fungus Cordyceps sinensis (Cs-HK1). EPS2BW was mainly composed of galactomannan with about 16% (w/w) protein and 50 kDa average molecular weight. The galactomannan part consisted of
Tianwei Hao et al.
Water research, 71, 74-84 (2015-01-21)
The SANI(®) process (Sulfate reduction, Autotrophic denitrification and Nitrification Integrated) is a treatment system with low energy demands. The major bioreactor of this new technology is a sulfate-reducing up-flow sludge bed (SRUSB) that converts organics and provides electron donors for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service