15523-M
Glycerol
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Synonym(s):
1,2,3-Propanetriol, Glycerin
About This Item
99.0-101.0% (calc. on H2O free substance)
electrophoresis: suitable
Recommended Products
Agency
meets analytical specifications of BP
meets analytical specifications of E422
meets analytical specifications of Ph. Eur.
meets analytical specifications of USP
Quality Level
reg. compliance
meets analytical specifications of FCC
vapor density
3.1 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
grade
puriss.
Assay
99.0-101.0% (alkalimetric)
99.0-101.0% (calc. on H2O free substance)
form
liquid
autoignition temp.
698 °F
quality
meets analytical specification of Ph. Eur., BP, USP, FCC, E422
technique(s)
IR spectroscopy: suitable
electrophoresis: suitable
impurities
acidity or alkalinity, complies
acrolein, glucose and NH4-compounds, complies
ester, complies
fatty acids and esters, complies
impurities A and related substances (GC), complies
residual solvents, complies
sugar, complies
≤0.003% free acid (as CH3COOH)
≤0.2% 1,2,4-butantriole (GC)
≤10 ppm aldehydes (as HCHO)
≤2% water (Karl Fischer)
≤30 ppm halogen compounds (as Cl)
≤5 ppm heavy metals (as Pb)
ign. residue
≤0.01% (as SO4)
refractive index
n20/D 1.471-1.474
n20/D 1.474 (lit.)
pH
5.5-8
bp
182 °C/20 mmHg (lit.)
mp
20 °C (lit.)
density
1.2580-1.2644 g/mL at 20 °C
≥1.257 g/mL at 25 °C
1.25 g/mL (lit.)
SMILES string
OCC(O)CO
anion traces
chloride (Cl-): ≤10 ppm
sulfate (SO42-): ≤10 ppm
cation traces
As: ≤1 ppm
Cu: ≤0.001%
Pb: ≤1 ppm
Zn: ≤0.001%
suitability
complies for appearance of solution
complies for reaction against H2SO4
corresponds for chromatography
passes test for identity (IR)
InChI
1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2
InChI key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Biochem/physiol Actions
- a supplement during cell culture of Mycobacterium tuberculosis and Mycobacterium avium.
- a fuel during designing enzymatic biofuel cell.
- a liquid composite matrix with 4-HCCA and 3-aminoquinoline for analysis of neutral and acidic glycans.
- a matrix for fast atom bombardment MS.
- may be employed as liquid matrix for the quantification studies by MALDI (Matrix-assisted laser desorption/ionization mass spectrometry) analysis.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
320.0 °F - closed cup
Flash Point(C)
160 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service