Skip to Content
Merck
All Photos(1)

Key Documents

8.18705

Sigma-Aldrich

Chloramine T trihydrate

for synthesis

Synonym(s):

Chloramine T trihydrate, Tosylchloramide sodium, N-Chloro-4-toluenesulfonamide sodium salt, Chlorine bleaching solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClNNaO2S · 3H2O
CAS Number:
Molecular Weight:
281.69
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-854-7
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

solid

potency

935 mg/kg LD50, oral (Rat)

pH

8-10 (20 °C, 50 g/L in H2O)

solubility

150 g/L

bulk density

540‑680 kg/m3

storage temp.

2-30°C

SMILES string

[S](=NCl)(=O)([O-])c1ccc(cc1)C

InChI

1S/C7H8ClNO2S/c1-6-2-4-7(5-3-6)12(10,11)9-8/h2-5H,1H3,(H,9,10,11)/p-1

InChI key

VQWVWLDBJISZHX-UHFFFAOYSA-M

Related Categories

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service