Skip to Content
Merck
All Photos(1)

Key Documents

8.14479

Sigma-Aldrich

Samarium

powder for synthesis

Synonym(s):

Samarium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
MDL number:
UNSPSC Code:
12141612
EC Index Number:
231-128-7
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

powder

mp

1072 °C

storage temp.

2-30°C

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Analysis Note

Assay (on Sm): ≥ 99.7 %
Identity (ICP-OES): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Water-react 3

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service