Skip to Content
Merck
All Photos(4)

Key Documents

1.04323

Millipore

modified Tryptic Soy Agar- Dehydrated Culture Media

NutriSelect® Prime, with Lecithin and Polysorbate 80, USP, EP, JP, EN 17141, ISO 21149, powder, Suitable for use in Halal and Kosher certified processes

Synonym(s):

TSA, soy agar, CASO agar, Casein-peptone Soymeal-peptone Agar, TSA medium, soybean casein digest agar, tryptic soy (caso) agar, tryptic soy agar dehydrated, Tryptone Soy Agar, Casein Soya Agar, Soybean Casein digest Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EN 17141
EP
ISO 21149
JP
USP

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.3 ± 0.2 (25.0 °C)

application(s)

cosmetics
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

The combination of the two peptones, enzymatic digest of casein and of soybean provides a high nutrition by supplying organic nitrogen, amino acids, and longer-chained peptides. In this complex medium the osmotic balance is supplied by sodium chloride whilst agar-agar is the solidifying agent.
This modification of Tryptic Soy Agar contains lecithin and polysorbate 80 (also known as Tween® 80) to inactivate residual disinfectants when sampling. EP, JP, USP, EN 17141 and EN ISO 21249 recommend incorporating lecithin to neutralize quaternary ammonium compounds (QACs), para-hydroxybenzoates (parabens) and bis-biguanides and to add polysorbate 80 to neutralize QACs, iodine and parabens. Polysorbate 80 is reported as well for neutralizing phenols, hexachlorophene, and formalin.
Tryptic Soy Agar (TSA) with Lecithin and Polysorbate 80 can be used to prepare plates for different usage, e.g. contact plates for surface-sampling, plates for surface-plating and air-sampling techniques, for membrane filtration and for pour-plating technique.

Application

NutriSelect® prime Tryptic Soy Agar halal with Lecithin and Polysorbate 80 acc. EP, USP, JP, EN 17141 and ISO 21149 is used for enumeration and detection of aerobic mesophilic microorganisms (Total aerobic microbial count) present in cosmetics, pharmaceutical and other products containing preservatives and for determining the sanitation efficiency of containers, equipment, and work area (environmental monitoring).

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service