1.00988
1-Butanol
EMPROVE® ESSENTIAL, ChP, NF
Pharma Manufacturing
Synonym(s):
1-Butanol, Butan-1-ol, n-Butyl alcohol, BuOH
About This Item
Recommended Products
Agency
ChP
NF
Quality Level
vapor pressure
6.7 hPa ( 20 °C)
product line
EMPROVE® ESSENTIAL
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
340 °C
expl. lim.
1.4-11.3 % (v/v)
technique(s)
API processing | polymorph screening: suitable
pH
7 (20 °C, 70 g/L in H2O)
bp
116-118 °C/1013 hPa
mp
-89 °C
transition temp
flash point 34 °C
density
0.81 g/cm3 at 20 °C
application(s)
liquid formulation
pharmaceutical
storage temp.
2-30°C
SMILES string
OCCCC
InChI
1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3
InChI key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F - Pensky-Martens closed cup
Flash Point(C)
35 °C - Pensky-Martens closed cup
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service