Skip to Content
Merck
All Photos(1)

Key Documents

1.00399

Supelco

N,N-Dimethylacetamide

for headspace gas chromatography SupraSolv®

Synonym(s):

N,N-Dimethylacetamide, Acetic acid dimethylamide, Acetyldimethylamine, DMAC, DMA, DMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-826-4
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
GC-HS: suitable
bp:
165-166 °C/1013 hPa
vapor pressure:
1.76 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

1.76 hPa ( 20 °C)

Quality Level

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

400 °C

potency

5830 mg/kg LD50, oral (Rat)

expl. lim.

1.7-11.5 % (v/v)

technique(s)

GC-HS: suitable

pH

4 (20 °C, 200 g/L in H2O)

bp

165-166 °C/1013 hPa

mp

-20 °C

transition temp

flash point 64 °C

solubility

soluble >1000 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(C)C(=O)C

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

General description

SupraSolv® high-purity solvents for headspace gas chromatography (HS-GC) are developed particularly for the analysis of residual solvents (RS) in drug substances, excipients and drug products according to ICH (International Council for Harmonizaton of Technical Requirements for Registration of Pharmaceuticals for Human Use) Guideline Q3C»Impurities: Guideline for Residual Solvents«, European Pharmacopeia (Ph. Eur. Chapter 2.4.24) and United States Pharmacopeia (USP Chapter 467).

Application

Suitable as a solvent for analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph. Eur. and USP:

  • Every residual solvent of class 1 acc. ICH ≤ 1 µg/g
  • Every residual solvent of class 2 acc. ICH ≤ 10 µg/g
  • Every residual solvent of class 3 acc. ICH ≤ 50 µg/g

Features and Benefits

  • high application reliability due to the application test performed on every batch produced
  • application-oriented quality due to specified concentrations of residual solvents as defined by ICH

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service