Skip to Content
Merck
All Photos(2)

Key Documents

07-571-I

Sigma-Aldrich

Anti-nNOS/NOS I Antibody

0.5 mg/mL, from rabbit

Synonym(s):

Nitric oxide synthase, brain, bNOS, Constitutive NOS, NC-NOS, NOS type I, Neuronal NOS, N-NOS, nNOS, Peptidyl-cysteine S-nitrosylase NOS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

concentration

0.5 mg/mL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nos1(18125)
rat ... Nos1(24598)

General description

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. nNOS/NOS I, also known as NC-NOS, or NOS type I, or Neuronal NOS, or Peptidyl-cysteine S-nitrosylase NOS1 and bNOS produces nitric oxide (NO) which is a messenger molecule with diverse functions throughout the body. In the brain and peripheral nervous system, NO displays many properties of a neurotransmitter. NOS I has been suggested to have nitrosylase activity and to mediate cysteine S-nitrosylation of cytoplasmic target proteins such SRR. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) & endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. Cytokines such as interferon-gamma (IFN), tumor necrosis factor (TNF), interleukin-1 and -2, and lipopolysaccarides (LPS) cause an increase in iNOS mRNA, protein, and activity levels. Protein kinase C-stimulating agents exhibit the same effect on iNOS activity. Human iNOS is regulated by calcium/calmodulin (in contrast with mouse NOS2).

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to the near N-terminus of rat nNOS/NOS I.

Application

Anti-nNOS/NOS I Antibody is an antibody against nNOS/NOS I for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected nNOS/NOS I in rat cerebellum and cerebral cortex tissues, and mouse hippocampus and thalamus tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in rat brain microsomal preparation.

Western Blotting Analysis: A 1:500 dilution of this antibody detected nNOS/NOS I in 10 µg of rat brain microsomal preparation.

Target description

~165 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Linkage

Replaces: 07-571

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arina Ponomarenko et al.
Marine drugs, 20(8) (2022-08-26)
At present, the study of the neurotropic activity of polyunsaturated fatty acid ethanolamides (N-acylethanolamines) is becoming increasingly important. N-docosahexaenoylethanolamine (synaptamide, DHEA) is a highly active metabolite of docosahexaenoic acid (DHA) with neuroprotective, synaptogenic, neuritogenic, and anti-inflammatory properties in the nervous
Shao-Rui Chen et al.
Neuropharmacology, 125, 156-165 (2017-07-30)
The role of nitric oxide (NO) in nociceptive transmission at the spinal cord level remains uncertain. Increased activity of spinal N-methyl-d-aspartate (NMDA) receptors contributes to development of chronic pain induced by peripheral nerve injury. In this study, we determined how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service