Skip to Content
Merck
All Photos(1)

Key Documents

P-100

Supelco

α-Pyrrolidinopropiophenone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO · HCl
CAS Number:
Molecular Weight:
239.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(C(C1=CC=CC=C1)=O)N2CCCC2.Cl

InChI

1S/C13H17NO.ClH/c1-11(14-9-5-6-10-14)13(15)12-7-3-2-4-8-12;/h2-4,7-8,11H,5-6,9-10H2,1H3;1H

InChI key

VQCZQKJXTMYEQV-UHFFFAOYSA-N

General description

α-Pyrrolidinopropiophenone hydrochloride is a stimulant drug, which affects the central nervous system.
Also known as α-PPP, this designer drug is a substituted cathinone found in bath salt blends and imitation “ecstasy” tablets. α-PPP possesses stimulant effects and is similar in structure to the synthetic cathinones MDPV and pyrovalerone. This Certified Spiking Solution® is suitable as starting material in calibrators and controls for synthetic cathinone testing methods by LC/MS or GC/MS in clinical toxicology, urine drug testing, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

a-PVP (?flakka?): a new synthetic cathinone invades the drug arena
Katselou M, et al.
Forensic Toxicology, 34(1), 41-50 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service