Skip to Content
Merck
All Photos(2)

Key Documents

850200P

Avanti

Regadenoson

2-[4-[(methylamino)carbonyl]-1H-pyrazol-1-yl]adenosine, powder

Synonym(s):

Lexiscan; CVT-3146

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N8O5
CAS Number:
Molecular Weight:
390.35
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (850200P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850200P

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[n]4(ncc(c4)C(=O)NC)c1nc2[n](cnc2c(n1)N)[C@@H]3O[C@@H]([C@H]([C@H]3O)O)CO

InChI

1S/C15H18N8O5/c1-17-13(27)6-2-19-23(3-6)15-20-11(16)8-12(21-15)22(5-18-8)14-10(26)9(25)7(4-24)28-14/h2-3,5,7,9-10,14,24-26H,4H2,1H3,(H,17,27)(H2,16,20,21)/t7-,9-,10-,14-/m1/s1

InChI key

LZPZPHGJDAGEJZ-AKAIJSEGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Regadenoson is an adenosine analog.[1]

Biochem/physiol Actions

Regadenoson (Lexiscan) is an A2A adenosine receptor agonist used as a coronary vasodilator in pharmacologic stress testing. Recently, it has been demonstrated that regadenoson can temporarily disrupt the integrity of the blood-brain barrier (BBB) by inhibiting P-glycoprotein function.
Regadenoson can be used in patients with obstructive lung disease and renal impairment for myocardial perfusion imaging (MPI).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (850200P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel D Cerqueira
The American journal of cardiology, 94(2A), 33D-40D (2004-07-21)
Adenosine and dipyridamole, the currently available vasodilators for myocardial perfusion imaging, produce hyperemic coronary flow by stimulating A(2A) adenosine receptors on arteriolar vascular smooth muscle cells. However, both vasodilators nonselectively activate A(1), A(2B), and A(3) adenosine receptors, which contributes to
G M Bengalorkar et al.
Journal of postgraduate medicine, 58(2), 140-146 (2012-06-22)
Single-photon emission computerized tomography for myocardial perfusion imaging (MPI) is a non-invasive technique. MPI is performed by subjecting the patient to exercise or by using a pharmacological stress agent. Regadenoson is a selective A 2A adenosine receptor agonist used when
Do-Geun Kim et al.
The Journal of clinical investigation, 126(5), 1717-1733 (2016-04-05)
The blood-brain barrier (BBB) protects the brain from toxic substances within the peripheral circulation. It maintains brain homeostasis and is a hurdle for drug delivery to the CNS to treat neurodegenerative diseases, including Alzheimer's disease and brain tumors. The drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service