Skip to Content
Merck
All Photos(2)

Key Documents

850100P

Avanti

18:1 Lyso PI

1-oleoyl-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt), powder

Synonym(s):

1-(9Z-octadecenoyl)-2-hydroxy-sn-glycero-3-phosphoinositol (ammonium salt); PI(18:1(9Z)/0:0); 110720

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H54NO12P
CAS Number:
Molecular Weight:
615.69
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPI; may contain up to 10% of the 2-LPI isomer, TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850100P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850100P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850100P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

General description

Phosphatidylinositol (PtdIns) is a structural phospholipid. PI is synthesized by mycobacteria and other bacterial genera in prokaryotes.

Application

18:1 Lyso PI is suitable for use to quantify the fatty acids, lysophospholipids (LPLs), and phospholipids (PLs) and LPAs in gintonin-enriched fraction (GEF) by liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Biochem/physiol Actions

Phosphatidylinositol (PI) participates in polyphosphoinositide turnover. It can be used as a substrate to produce several molecules, that can modulate various cellular functions. In eukaryotes, phosphatidylinositol (PI)-based lipids (phosphoinositides) plays a key role in intracellular signalling. It is also involved in the anchoring of glycosylphosphatidylinositol-linked proteins to the membrane.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850100P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850100P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Phosphatidylinositol 4-Kinase from Bovine Brain
Methods in Neurosciences, 18, 93-99 (1993)
Bioactive lipids in gintonin-enriched fraction from ginseng
Cho HJ, et al.
Journal of ginseng research (2017)
Phosphatidylinositol Turnover and Receptors
reference module in biomedical sciences (2016)
Structural basis for phosphatidylinositol-phosphate biosynthesis
Clarke OB, et al.
Nature Communications, 6, 8505-8505 (2015)
Qingxin Shi et al.
Food & function, 10(12), 7782-7792 (2019-11-30)
Triterpenoid saponins from Kuding tea have demonstrated preventive effects on hyperlipidaemia induced by a high-fat diet. Lysoglycerophospholipids (Lyso-GPLs) are known to be associated with proatherogenic conditions such as hyperlipidaemia. In this study, a target profiling strategy based on a multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service