Skip to Content
Merck
All Photos(1)

Key Documents

790310P

Avanti

Transfection Reagent I

Avanti Research - A Croda Brand 790310P, powder

Synonym(s):

Transfection Reagent I, Transfection Reagent I, contains DOTAP:DOPE (1:1 w/w)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (790310P-10mg)
pkg of 2 × 100 mg (790310P-200mg)
pkg of 5 × 100 mg (790310P-500mg)
pkg of 1 × 25 mg (790310P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790310P

application(s)

advanced drug delivery

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

General description

Transfection Reagent I comprises the cationic lipid 1,2-dioleoyloxypropyl)-N,N,N-trimethylammonium chloride (DOTAP) and the neutral lipid DOPE (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine. DOTAP is a surfactant with double chain quaternary ammonium. The DOTAP/DOPE liposome takes lamellar and inverted hexagonal structures.

Application

Transfection Reagent I has been used in the preparation of liposomes along with cholesterol for fluorescence-based live-cell imaging studies in the mouse embryonic fibroblasts cells. It may be used for the transfection of DNAzyme (Dz13) into veins.

Biochem/physiol Actions

Transfection Reagent I is used in large unilamellar cationic vesicles (LUVs) preparation and lipoplex generation. It improves transfection efficiency. DOTAP/DOPE (1,2-dioleoyloxypropyl)-N,N,N-trimethylammonium chloride /(1,2-dioleoyl-sn-glycero-3-phosphoethanolamine) is safe transfection agent for human administration. It inhibits the blood vessel thickening and is a promising candidate for use in coronary artery bypass grafting (CABG).

Packaging

5 mL Clear Glass Sealed Ampule (790310P-10mg)
5 mL Clear Glass Sealed Ampule (790310P-200mg)
5 mL Clear Glass Sealed Ampule (790310P-25mg)
5 mL Clear Glass Sealed Ampule (790310P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autophagy and formation of tubulovesicular autophagosomes provide a barrier against nonviral gene delivery
Roberts R, et al.
Autophagy, 9(5), 667-682 (2013)
Inhibition of vein graft stenosis with a c-jun targeting DNAzyme in a cationic liposomal formulation containing 1, 2-dioleoyl-3-trimethylammonium propane (DOTAP)/1, 2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE)
Li Y, et al.
International Journal of Cardiology, 168(4), 3659-3664 (2013)
Yue Li et al.
International journal of cardiology, 168(4), 3659-3664 (2013-07-28)
Coronary artery bypass grafting (CABG) is among the most commonly performed heart surgical procedures. Saphenous vein graft failure due to stenosis impedes the longer-term success of CABG. A key cellular event in the process of vein graft stenosis is smooth
Bieong-Kil Kim et al.
Biochimica et biophysica acta, 1848(10 Pt A), 1996-2001 (2015-06-27)
The effects of lipid compositions on their physicochemical properties and transfection efficiencies were investigated. Four liposome formulations with different 1,2-dioleoyl-3-trimethylammoniumpropane (DOTAP) to dioleoylphosphatidylethanolamine (DOPE) weight ratios were investigated, that is, weight ratios 1:0 (T1P0), 3:1 (T3P1), 1:1 (T1P1), and 1:3
DOTAP/DOPE and DC-Chol/DOPE lipoplexes for gene delivery: zeta potential measurements and electron spin resonance spectra
Ciani L, et al.
Biochimica et Biophysica Acta - Biomembranes, 1664(1), 70-79 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service