Key Documents
SLW1594/02D
Pyrex® Vacuum desiccator
Metal perforated plate not included; part number SLW1597/02D
Synonym(s):
Pyrex, SciLabware 1594/02D
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
feature
Metal perforated plate not included; part number SLW1597/02D
packaging
pkg of 1 ea
manufacturer/tradename
SciLabware 1594/02D
SciLabware
flange I.D.
220 mm
flange Overall H
120 mm , Height from ledge to flange
General description
Glass vacuum desiccator
  • Designed for use under vacuum
  • Sturdy construction in borosilicate glass
  • Ground glass flanges
  • Cover has a 24/29 socket to accept desiccator stopcocks
  • Plates are positioned on an internal ledge within the base
Metal perforated plate not included; part number SLW1597/02D
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)