Skip to Content
Merck
All Photos(1)

Key Documents

QBD10379

Sigma-Aldrich

SPDP-dPEG®24-NHS ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H113N3O29S2
Molecular Weight:
1440.70
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCSSC1=NC=CC=C1)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON2C(CCC2=O)=O)=O

Features and Benefits

SPDP-dPEG®24-NHS ester has an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a sulfhydryl-reactive 2-pyridyldithiol group at the opposite end. This has a single molecular weight PEG spacer arm, which confers greater water solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Protein aggregation and precipitation also are greatly reduced with SPDP-dPEG crosslinkers compared to crosslinkers that have only hydrocarbon spacers. dPEG spacer of specific length (97.3 Å) provides precise distance control between the SPDP moiety and the target molecule. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using SPDP reagents.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service