Skip to Content
Merck
All Photos(3)

Documents

P28263

Sigma-Aldrich

2-Phenylphenol

99%

Synonym(s):

2-Hydroxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4OH
CAS Number:
Molecular Weight:
170.21
Beilstein:
606907
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7 mmHg ( 140 °C)

Assay

99%

form

solid

bp

282 °C (lit.)

mp

57-59 °C (lit.)

fluorescence

λex 231 nm; λem 356 nm

SMILES string

Oc1ccccc1-c2ccccc2

InChI

1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H

InChI key

LLEMOWNGBBNAJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenylphenol, also known as o-phenylphenol, is commonly used as a chemical intermediate for the synthesis of pharmaceuticals, dyes, and fragrances.

Application

2-Phenylphenol can be used as a precursor in the synthesis of:
  • Dibenzofurans via palladium-catalyzed, phenol-directed C-H activation/C-O cyclization or copper-catalyzed aerobic C-H activation, followed by cycloetherification.
  • 6H-Dibenzo[b,d]pyran-6-ones via ruthenium-catalyzed carbonylative C-H cyclization.
  • o-Phenylenes, a class of conjugated oligomers via an iterative sequence of Suzuki-Miyaura coupling reactions.
  • o,o,p-Oligophenylenes via palladium-catalyzed C-H arylation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-Catalyzed Carbonylative C?H Cyclization of 2-Arylphenols: A Novel Synthetic Route to 6 H-Dibenzo [b, d] pyran-6-ones.
Inamoto K, et al.
Organic Letters, 15(15), 3962-3965 (2013)
Cu-catalyzed oxidative C (sp2)-H cycloetherification of o-arylphenols for the preparation of dibenzofurans.
Zhao J, et al.
Organic Letters, 14(4), 1078-1081 (2012)
Synthesis of dibenzofurans via Palladium-catalyzed phenol-directed C?H activation/C?O cyclization.
Xiao B, et al.
Journal of the American Chemical Society, 2011, 9250-9253 (2011)
Parent o-phenylene oligomers: synthesis, conformational behavior, and characterization.
Mathew S M and Hartley C S
Macromolecules, 44(21), 8425-8432 (2011)
Repetitive synthetic method for o, o, p-oligophenylenes using C?H arylation.
Manabe K and Kimura T
Organic Letters, 15(2), 374-377 (2013)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service