E45600
Ethyl 1-piperazinecarboxylate
99%
Synonym(s):
1-Ethoxycarbonylpiperazine, Piperazine-1-carboxylic acid ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H14N2O2
CAS Number:
Molecular Weight:
158.20
Beilstein:
125780
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.477 (lit.)
bp
273 °C (lit.)
density
1.08 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)N1CCNCC1
InChI
1S/C7H14N2O2/c1-2-11-7(10)9-5-3-8-4-6-9/h8H,2-6H2,1H3
InChI key
LNOQURRKNJKKBU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Natalia Szkaradek et al.
Anti-cancer agents in medicinal chemistry, 19(16), 1949-1965 (2019-04-06)
Natural plant metabolites and their semisynthetic derivatives have been used for years in cancer therapy. Xanthones are oxygenated heterocyclic compounds produced as secondary metabolites by higher plants, fungi or lichens. Xanthone core may serve as a template in the synthesis
Karolina Klesiewicz et al.
The Journal of antibiotics, 69(11), 825-834 (2016-03-31)
A series of 20 xanthone derivatives was synthesized and evaluated for anti-Helicobacter pylori (H. pylori) activity. Qualitative and quantitative in vitro tests using the Kirby-Bauer method (agar disc-diffusion method) were performed. The tested compounds were screened against clarithromycin- and/or metronidazole-resistant
Paul J Shami et al.
Journal of medicinal chemistry, 49(14), 4356-4366 (2006-07-11)
The literature provides evidence that metabolic nitric oxide (NO) release mediates the cytotoxic activities (against human leukemia and prostate cancer xenografts in mice) of JS-K, a compound of structure R(2)N-N(O)=NO-Ar for which R(2)N is 4-(ethoxycarbonyl)piperazin-1-yl and Ar is 2,4-dinitrophenyl. Here
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service