Skip to Content
Merck
All Photos(1)

Key Documents

DWK73805B18415

Kimble® Closed Top Linerless Polypropylene Screw Thread Cap

18-415, natural screw cap

Synonym(s):

73805B-18415

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.17

material

natural screw cap
red polypropylene screw cap (Push fit)

packaging

pkg of 1 packs

manufacturer/tradename

Kimble® 73805B-18415

fitting

thread

Looking for similar products? Visit Product Comparison Guide

Application

Closure solution for vials and bottles

Features and Benefits

  • Economical, one-piece construction
  • Unique design provides exceptional sealing properties
  • Autoclavable
  • Designed from ASTM Specification E982

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service