Skip to Content
Merck
All Photos(1)

Key Documents

DWK217217109

Duran® Round Bottom Flask

narrow-mouth neck, WITH BEADED RIM

Synonym(s):

217217109

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

narrow-mouth neck
round bottom

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 217217109

capacity

4000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Uniform wall thickness distribution makes these flasks ideal for heating applications. Heating Size 2000, 3000, 5000 ml non-DIN EN ISO size., sizes 12000 ml and 20000 ml not DIN ISO but they are ASTM E1403 Boro 3.3

Features and Benefits

The geometry permits very uniform heating. Fired-on,

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service