Skip to Content
Merck
All Photos(1)

Key Documents

BMSDP0150

Benchmark Pulse 150 Ultrasonic Homogenizer with 6mm horn and soundproof box

AC/DC input 115 V

Synonym(s):

bead homogenization, bead grinder,cell lysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101504
NACRES:
NB.13

feature

Includes 6mm horn and soundproof box

manufacturer/tradename

Benchmark Scientific DP0150

Looking for similar products? Visit Product Comparison Guide

General description

The Pulse 150 Ultrasonic Homogenizer uses cavitation and ultrasonic waves to break cells in suspension, shear DNA and prepare samples for ChIP assays. With up to 150 watts of power, the unit processes samples from0.1 ml to 150 ml (with the appropriate horn).

All components needed for homogenization are included with the Pulse 150 a controller, transducer, temperature probe, 6 mm diameter titanium alloy horn and wrenches to change horns. A soundproof box with sample platform is also supplied as standard. The box reduces the noise generated by the sonicator. The 6mm horn is appropriate for volumes from 10ml to 100ml. Optional horns are available for other volumes.

Operating parameters are set and viewed on the 4.3 inch color touch screen. Pulsed operation is easily programmed as is the power level. Twenty programs can be stored in memory and recalled as needed. Alarms alert the user to over temperature, over/no load and expiration of time.

Application

Ultrasonic homogenization and lysis

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service