Skip to Content
Merck
All Photos(1)

Documents

765759

Sigma-Aldrich

3-(Trifluoromethyl)-1H-indene

95%

Synonym(s):

3-(Trifluoromethyl)indene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F3
CAS Number:
Molecular Weight:
184.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.489

storage temp.

2-8°C

SMILES string

FC(F)(F)C1=CCc2ccccc12

InChI

1S/C10H7F3/c11-10(12,13)9-6-5-7-3-1-2-4-8(7)9/h1-4,6H,5H2

InChI key

PCPRMODISMAALW-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service