Skip to Content
Merck
All Photos(2)

Key Documents

753130

Sigma-Aldrich

Barium bis(trifluoromethanesulfonimide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4BaF12N2O8S4
CAS Number:
Molecular Weight:
697.62
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: barium
reagent type: catalyst

mp

>300 °C

SMILES string

FC(F)(F)S(=O)(=O)N([Ba]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/2C2F6NO4S2.Ba/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q2*-1;+2

InChI key

ZFICMXSMTNWQDX-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service