Skip to Content
Merck
All Photos(1)

Key Documents

697869

Sigma-Aldrich

Methyl 2-fluoropyridine-4-carboxylate

97%

Synonym(s):

Methyl 2-Fluoroisonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6FNO2
CAS Number:
Molecular Weight:
155.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.488

density

1.251 g/mL at 25 °C

SMILES string

COC(=O)c1ccnc(F)c1

InChI

1S/C7H6FNO2/c1-11-7(10)5-2-3-9-6(8)4-5/h2-4H,1H3

InChI key

MTZKFHSYUSUUIU-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service