Skip to Content
Merck
All Photos(1)

Key Documents

279889

Sigma-Aldrich

(S)-(+)-2-Phenylbutyric acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(C6H5)CO2H
CAS Number:
Molecular Weight:
164.20
Beilstein:
2045208
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]19/D +92°, c = 0.9 in toluene

refractive index

n20/D 1.516 (lit.)

bp

98-100 °C/0.3 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CC[C@H](C(O)=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)/t9-/m0/s1

InChI key

OFJWFSNDPCAWDK-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block. Resolving agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service