Skip to Content
Merck
All Photos(2)

Key Documents

258040

Sigma-Aldrich

3,4-Dimethoxybenzoyl chloride

98%

Synonym(s):

Veratroyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCl
CAS Number:
Molecular Weight:
200.62
Beilstein:
783596
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

70-73 °C (lit.)

SMILES string

COc1ccc(cc1OC)C(Cl)=O

InChI

1S/C9H9ClO3/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3

InChI key

VIOBGCWEHLRBEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dimethoxybenzoyl chloride has been used in synthesis of:
  • (+)-5-(3,4-dimethoxyphenyl)-4-[[N-[(4S)-2-oxo-4-(phenylmethyl)-2-oxazolidinyl]]carbonyl] oxazole and its enantiomer
  • N-(2,5-dibromophenyl)-3,4-dimethoxybenzamide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian S. Tang et al.
The Journal of organic chemistry, 61(25), 8750-8754 (1996-12-13)
Both (4S-(+)-3-(isocyanoacetyl)-4-(phenylmethyl)-2-oxazolidinone (R)-1 and its enantiomer (S)-1 have been synthesized as potentially useful synthons in asymmetric synthesis. Optically active (+)-5-(3,4-dimethoxyphenyl)-4-[[N-[(4S)-2-oxo-4-(phenylmethyl)-2-oxazolidinyl]]carbonyl] oxazole (S)-2 and its enantiomer (R)-2 obtained by treating 3,4-dimethoxybenzoyl chloride with (S)-1 and (R)-1, respectively, in the presence of
Catriona G Mortimer et al.
Journal of medicinal chemistry, 49(1), 179-185 (2006-01-06)
A series of new 2-phenylbenzothiazoles has been synthesized on the basis of the discovery of the potent and selective in vitro antitumor properties of 2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole (8n; GW 610, NSC 721648). Synthesis of analogues substituted in the benzothiazole ring was achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service